Order Entry
India
ContactUsLinkComponent
2-Propanol ≥70% in aqueous solution, AR®, Macron Fine Chemicals™
Flammable
Irritant
2-Propanol ≥70% in aqueous solution, AR®, Macron Fine Chemicals™
Certificates
Flammable
Irritant
2-Propanol ≥70% in aqueous solution, AR®, Macron Fine Chemicals™
Certificates

Specification Test Results

  • Specific Gravity at 20°/20°C:
    0.872 - 0.883
  • Acidity (mL):
    ≤1.0
  • Nonvolatile Residue:
    ≤0.01 %
  • Assay (by GC):
    68.0 - 72.0 %

Specification Test Results

Related Information

Frequently Bought Together