


Tetrahydrofuran ≥99.0% stabilised, J.T.Baker®
Catalog # V530-01
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Steel pail
- Pk:20 L
- Seq:5302
Specifications
Specification Test Results
- Assay (C₄H₈O) (by GC, corrected for water):≥99 %
Specification Test Results
About this item
:
:72.11 g/mol
Merck Index:12,09351
Boiling Pt:66 °C (1013 hPa)
Melting Pt:–108,5 °C
Density:0,888 g/cm³ (20 °C)
Flash Pt:–21,5 °C
ADR:3,II
:Management of Change (MOC) category = N
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
Storage Temperature:Ambient
EINECS:203-726-8
UN:2056
REACH:05-2114615188-48
:GHS02
:GHS08
:GHS07
CAS Number:109-99-9
Formula:C₄H₈O
MDL Number:MFCD00005356