Order Entry
India
ContactUsLinkComponent
EDTA dipotassium salt dihydrate 99.0-101.0%, BAKER ANALYZED® Electronic Grade, J.T.Baker®
EDTA dipotassium salt dihydrate 99.0-101.0%, BAKER ANALYZED® Electronic Grade, J.T.Baker®
Supplier:  Avantor
Certificates
EDTA dipotassium salt dihydrate 99.0-101.0%, BAKER ANALYZED® Electronic Grade, J.T.Baker®
Supplier:  Avantor

Specification Test Results

  • Assay (C₁₀H₁₄K₂N₂O₈·2H₂0):
    99.0 - 101.0 %
  • Insoluble Matter:
    ≤0.005 %
  • pH of 5% Solution:
    4.0 - 6.0
  • Chloride (Cl):
    ≤100 ppm
  • Iron (Fe):
    ≤100 ppm
  • Lead (Pb):
    ≤50 ppm
  • Sodium (Na):
    ≤1500.000 ppm

Specification Test Results

Frequently Bought Together