

Sodium hydroxide 10.0 N in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Boiling Pt:119…135 °C (1013 hPa)
Melting Pt:9…10 °C
Density:1,53 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
ADR:8,II
:Management of Change (MOC) category = NI
UN:1824
:GHS05
:GHS07
Specification Test Results
- Normality:9.95 - 10.05
- Identification:Passes Test
Specification Test Results
Frequently Bought Together