2,3,5-Triphenyltetrazolium chloride (TTC) 90-100%, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:298-96-4
:
Formula:C₁₉H₁₅ClN₄
MDL Number:MFCD00011963
Merck Index:13,09815
Boiling Pt:∼360 °C (1013 hPa)
Melting Pt:235 °C
:Management of Change (MOC) category = N
:334.81 g/mol
EINECS:206-071-6
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Identification (by IR):Passes Test
- Loss on Drying at 105°C:≤5.0 %
- Residue after Ignition:Passes Test
- Sensitivity:Passes Test
- Solubility:Passes Test
Specification Test Results
Frequently Bought Together