Order Entry
India
ContactUsLinkComponent
VWR® Traceable® Replacement Platinum RTD Stainless-Steel Probe
VWR® Traceable® Replacement Platinum RTD Stainless-Steel Probe
Catalog # 620-2810
Supplier:  VWR Collection
undefined
VWR® Traceable® Replacement Platinum RTD Stainless-Steel Probe
Catalog # 620-2810
Supplier:  VWR Collection
Specifications
  • Range:
    ‒100...200 °C
  • Description:
    VWR® Traceable® replacement platinum RTD stainless-steel probe for RTD thermometer (VWRI620-1981)
  • Length:
    10' PTFE cable
  • Environmentally Preferable:
  • Pk:
    1
  • Type:
    Platinum RTD
Specifications
About this item
Optimise performance of thermometer.
  • Replace the probe or keep a convenient spare
Replacement probe for RTD thermometer (VWRI620-1981).