Order Entry
India
ContactUsLinkComponent
 
 
 
Zinc acetate dihydrate ≥98%, granular, AR® ACS, Macron Fine Chemicals™
 
 
 
Zinc acetate dihydrate ≥98%, granular, AR® ACS, Macron Fine Chemicals™
 

 

  • ACS - Assay ((CH₃COO)₂Zn·2H₂O):
    98.0 - 101.0 %
  • Solubility:
    Passes Test
  • ACS - Chloride (Cl):
    ≤0.0005 %
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - Iron (Fe):
    ≤0.0005 %
  • ACS - Lead (Pb):
    ≤0.002 %
  • ACS - Sulfate (SO₄):
    ≤0.002 %
  • ACS - pH of 5% Solution at 25°C:
    6.0 - 7.0
  • ACS - Calcium (Ca):
    ≤0.005 %
  • ACS - Magnesium (Mg):
    ≤0.005 %
  • ACS - Potassium (K):
    ≤0.01 %
  • ACS - Sodium (Na):
    ≤0.05 %

 

 

Frequently Bought Together