


Nitric acid 68,0 - 70,0%, AR Select® ACS for trace metal analysis, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:7697-37-2
:
Formula:HNO₃
MDL Number:MFCD00011349
:63.01 g/mol
Merck Index:13,06608
Boiling Pt:120,5 °C (1013 hPa)
Melting Pt:–42 °C
Density:1,4 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2031
ADR:8,II
:Management of Change (MOC) category = NI
EINECS:231-714-2
:GHS03
:GHS05
:GHS07
Specification Test Results
- ACS - Assay (HNO₃):68.0 - 70.0 %
- Appearance:Passes Test
- ACS - Color (APHA):≤ 10
- ACS - Arsenic (As):≤ 0.010 ppm
- ACS - Arsenic and Antimony (as As):≤ 0.005 ppm
- ACS - Chloride (Cl):≤ 0.04 ppm
- ACS - Heavy Metals (as Pb):≤ 0.05 ppm
- ACS - Residue after Ignition:≤ 5 ppm
- ACS - Sulfate (SO₄):≤ 0.4 ppm
- Determine by Flame Photometry & ICP - Aluminum (Al):≤ 50.0 ppb
- Determine by Flame Photometry & ICP - Barium (Ba):≤ 1.0 ppb
- Determine by Flame Photometry & ICP - Beryllium (Be):≤ 1.0 ppb
- Determine by Flame Photometry & ICP - Bismuth (Bi):≤ 10 ppb
- Determine by Flame Photometry & ICP - Boron (B):≤ 10.0 ppb
- Determine by Flame Photometry & ICP - Cadmium (Cd):≤ 1.0 ppb
- Determine by Flame Photometry & ICP - * Calcium (Ca):≤ 200.0 ppb
- Determine by Flame Photometry & ICP - Chromium (Cr):≤ 10.0 ppb
- Determine by Flame Photometry & ICP - Cobalt (Co):≤ 5.0 ppb
- Determine by Flame Photometry & ICP - Copper (Cu):≤ 1.0 ppb
- Determine by Flame Photometry & ICP - Gallium (Ga):≤ 20.0 ppb
- Determine by Flame Photometry & ICP - Germanium (Ge):≤ 10.0 ppb
- Determine by Flame Photometry & ICP - Gold (Au):≤ 10.0 ppb
- Determine by Flame Photometry & ICP - Iron (Fe):≤ 40.0 ppb
- Determine by Flame Photometry & ICP - Lead (Pb):≤ 1.0 ppb
- Determine by Flame Photometry & ICP - Lithium (Li):≤ 5.0 ppb
- Determine by Flame Photometry & ICP - Magnesium (Mg):≤ 10.0 ppb
- Determine by Flame Photometry & ICP - Manganese (Mn):≤ 1.0 ppb
- Determine by Flame Photometry & ICP - Mercury (Hg):≤ 1.0 ppb
- Determine by Flame Photometry & ICP - Molybdenum (Mo):≤ 5.0 ppb
- Determine by Flame Photometry & ICP - Nickel (Ni):≤ 5.0 ppb
- Determine by Flame Photometry & ICP - Potassium (K):≤ 20.0 ppb
- Determine by Flame Photometry & ICP - * Silicon (Si):≤ 50.0 ppb
- Determine by Flame Photometry & ICP - Silver (Ag):≤ 5.0 ppb
- Determine by Flame Photometry & ICP - * Sodium (Na):≤ 500.0 ppb
- Determine by Flame Photometry & ICP - Strontium (Sr):≤ 1.0 ppb
- Determine by Flame Photometry & ICP - Thallium (Tl):≤ 20.0 ppb
- Determine by Flame Photometry & ICP - Tin (Sn):≤ 10.0 ppb
- Determine by Flame Photometry & ICP - Vanadium (V):≤ 1.0 ppb
- Determine by Flame Photometry & ICP - Zinc (Zn):≤ 5.0 ppb
- Determine by Flame Photometry & ICP - Zirconium (Zr):≤ 10 ppb
Specification Test Results
Frequently Bought Together