

Sodium cyanide ≥95%, granular, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:143-33-9
:
Formula:CNNa
MDL Number:MFCD00003523
Merck Index:13,08679
Boiling Pt:1496 °C (1013 hPa)
Melting Pt:563,9 °C
Density:1,595 g/cm³ (20 °C)
EINECS:205-599-4
UN:1689
ADR:6.1,I
:Management of Change (MOC) category = N
:49.01 g/mol
:GHS06
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Assay (NaCN) (by Ag titrn):≥ 95.0 %
- Chloride (Cl):≤ 0.15 %
- Phosphate (PO₄):≤ 0.02 %
- Sulfate (SO₄):≤ 0.05 %
- Sulfide (S):≤ 0.005 %
- Thiocyanate (SCN):≤ 0.02 %
- Iron (Fe):≤ 0.005 %
- Trace Impurities - Lead (Pb):≤ 5 ppm
Specification Test Results
Related Information
Certificates
Search by Lot NumberChemical Mining Supplies
Product Literature
spec_BAKR3662-01
Product specification
Frequently Bought Together