Order Entry
India
ContactUsLinkComponent
Trichloroacetic acid (TCA) ≥99.0% (by acidimetry on anhydrous substance), crystals, AR® ACS, Macron Fine Chemicals™
Corrosive
Irritant
Environment
Health
Trichloroacetic acid (TCA) ≥99.0% (by acidimetry on anhydrous substance), crystals, AR® ACS, Macron Fine Chemicals™
Certificates
undefined
Corrosive
Irritant
Environment
Health
Trichloroacetic acid (TCA) ≥99.0% (by acidimetry on anhydrous substance), crystals, AR® ACS, Macron Fine Chemicals™
Certificates

Specification Test Results

  • ACS - Assay (Cl₃CCOOH) (dried basis)(by acidimetry):
    ≥99.0 %
  • ACS - Clarity of Solution:
    Passes Test
  • ACS - Insoluble Matter:
    ≤0.01 %
  • ACS - Residue after Ignition:
    ≤0.03 %
  • ACS - Chloride (Cl):
    ≤0.002 %
  • ACS - Nitrate (NO₃):
    ≤0.002 %
  • ACS - Phosphate (PO₄):
    ≤5 ppm
  • ACS - Sulfate (SO₄):
    ≤0.02 %
  • ACS - Heavy Metals (as Pb):
    ≤0.002 %
  • ACS - Iron (Fe):
    ≤0.001 %
  • ACS - Substances Darkened by H₂SO₄:
    Passes Test

Specification Test Results

Related Information

Frequently Bought Together