Order Entry
India
ContactUsLinkComponent
 
 
Sodium hydroxide 5 N 97.5-100.5% Standard of Eli Lilly NF, ACS, Macron Fine Chemicals™
 
undefined
 
Sodium hydroxide 5 N 97.5-100.5% Standard of Eli Lilly NF, ACS, Macron Fine Chemicals™
 

 

  • NF - Total Alkali:
    97.5 - 100.5 %
  • NF - Carbonate (as Na₂CO₃):
    ≤3.0 %
  • NF - Content of Sodium:
    54.0 - 59.8 %
  • NF - Identification A (Sodium):
    Passes Test
  • NF - Identification B (pH):
    ≥11.0
  • NF - Insoluble Substances and Organic Matter:
    Passes Test
  • NF - Potassium (K):
    ≤0.5 %
  • ACS - Assay (NaOH):
    ≥97.0 %
  • ACS - Sodium Carbonate (Na₂CO₃):
    ≤1.0 %
  • ACS - Sulfate (SO₄):
    ≤0.003 %
  • ACS - Chloride (Cl):
    ≤0.005 %
  • ACS - Nitrogen Compounds (as N):
    ≤0.001 %
  • ACS - Phosphate (PO₄):
    ≤0.001 %
  • ACS - Heavy Metals (as Ag):
    ≤0.002 %
  • ACS - Iron (Fe):
    ≤0.001 %
  • ACS - Nickel (Ni):
    ≤0.001 %
  • ACS - Mercury (Hg)(by AAS):
    ≤0.100 ppm
  • ACS - Calcium (Ca):
    ≤0.005 %
  • ACS - Magnesium (Mg):
    ≤0.002 %
  • ACS - Potassium (K):
    ≤0.02 %

 

 

Frequently Bought Together