Order Entry
India
ContactUsLinkComponent
Orthophosphoric acid 85%, BAKER ANALYZED® ACS, J.T.Baker®
 
 
Orthophosphoric acid 85%, BAKER ANALYZED® ACS, J.T.Baker®
  0269-68
 :  Avantor
 :  
 
 
Orthophosphoric acid 85%, BAKER ANALYZED® ACS, J.T.Baker®
  0269-68
 :  Avantor
 :  

 

  • Pk:
    2,5 L
  • Seq:
    5096

 

 

  • ACS - Assay (H₃PO₄):
    ≥85.0 %
  • ACS - Antimony (Sb):
    ≤15.000 ppm
  • ACS - Arsenic (As):
    ≤1.000 ppm
  • ACS - Calcium (Ca):
    ≤0.002 %
  • ACS - Chloride (Cl):
    ≤3 ppm
  • ACS - Color (APHA):
    ≤10
  • ACS - Heavy Metals (as Pb):
    ≤10 ppm
  • ACS - Insoluble Matter:
    ≤0.001 %
  • ACS - Iron (Fe):
    ≤6 ppm
  • ACS - Magnesium (Mg):
    ≤0.002 %
  • ACS - Manganese (Mn):
    ≤0.500 ppm
  • ACS - Nitrate (NO₃):
    ≤5 ppm
  • ACS - Potassium (K):
    ≤6.000 ppm
  • ACS - Reducing Substances, mL:
    ≤1.0
  • ACS - Sodium (Na):
    ≤10.000 ppm
  • ACS - Sulfate (SO₄):
    ≤0.001 %
  • ACS - Volatile Acids (as CH₃COOH):
    ≤0.001 %

 

 

 :
 :98 g/mol
 :13,08662
 :28 °C
 :Ambient
 :231-633-2
 :1805
 :8,III
 :01-2119490790-32
 :GHS05
 :GHS07
 :7664-38-2
 :H₃PO₄
 :MFCD00011340