tri-Sodium citrate dihydrate 99.0-100.5% (by anhydrous basis), granular USP, FCC, J.T.Baker®
Catalog # 3649-B1
Supplier: Avantor
CAS Number:
Specifications
- Pk:10.098 g
- Seq:3905
Specifications
Specification Test Results
- USP - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis):99.0 - 100.5 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Identification C:Passes Test
- USP - Alkalinity:Passes Test
- USP - Water (H₂O):10.0 - 13.0 %
- USP - Tartrate:Passes Test
- FCC - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis):99.0 - 100.5 %
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- FCC - Alkalinity:Passes Test
- FCC - Lead (Pb):≤2 mg/kg
- FCC - Water (FCC):10.0 - 13.0 %
Specification Test Results
About this item
:
:294.1 g/mol
Merck Index:13,08675
Melting Pt:150 °C
Density:1,70 g/cm³
Storage Temperature:Ambient
EINECS:200-675-3
CAS Number:6132-04-3
Formula:Na₃C₆H₅O₇·2H₂O
MDL Number:MFCD00150031