Order Entry
India
ContactUsLinkComponent
Nickel(II) chloride solution (concentrate), J.T.Baker®
Irritant
Health
Nickel(II) chloride solution (concentrate), J.T.Baker®
Catalog # 3806-09
Supplier:  Avantor
CAS Number:  
Irritant
Health
Nickel(II) chloride solution (concentrate), J.T.Baker®
Catalog # 3806-09
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

Specification Test Results

  • Appearance (green solution):
    Passes Test
  • Assay (NiCl₂.6H₂O), g/L:
    100 - 140 g/l

Specification Test Results

About this item

 :
 :129.6 g/mol
 :Management of Change (MOC) category = NI
 :GHS07
 :GHS08
CAS Number:7718-54-9
Formula:Cl₂Ni
MDL Number:MFCD00011142