Sodium chloride 99.0-100.5% (dry basis), granular FCC, USP, J.T.Baker®
Catalog # 3628-X1
Supplier: Avantor
CAS Number:
Specifications
- Pk:1 KIT
- Seq:0075
Specifications
Specification Test Results
- Kit Components Meet Requirements:Passes Test
Specification Test Results
About this item
:
:58.44 g/mol
Merck Index:14,08599
Boiling Pt:1413 °C (1013 hPa)
Melting Pt:801 °C
Density:2,16 g/cm³
Storage Temperature:Ambient
EINECS:231-598-3
:GHS07
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477