D-(+)-Galactose kit ≥99% (by HPLC), Multi-Compendial low in endotoxins, J.T.Baker®
Catalog # 6322-X1
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Bag/kit
- Pk:1 KIT
- Seq:0075
Specifications
Specification Test Results
- Kit Components Meet Requirements:Passes Test
Specification Test Results
About this item
:
:180.16 g/mol
Density:1,72 g/cm³
:Management of Change (MOC) category = R
CAS Number:59-23-4
Formula:C₆H₁₂O₆
MDL Number:MFCD00151230