Order Entry
India
ContactUsLinkComponent
Potassium dihydrogen phosphate kit 98.0-100.5% (dried basis), crystals NF, Multi-Compendial, J.T.Baker®
Irritant
Potassium dihydrogen phosphate kit 98.0-100.5% (dried basis), crystals NF, Multi-Compendial, J.T.Baker®
Catalog # 3248-X3
Supplier:  Avantor
CAS Number:  
undefined
Irritant
Potassium dihydrogen phosphate kit 98.0-100.5% (dried basis), crystals NF, Multi-Compendial, J.T.Baker®
Catalog # 3248-X3
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Bag
  • Pk:
    1 KIT
  • Seq:
    0075

Specifications

Specification Test Results

  • Kit Components Meet Requirements:
    Passes Test

Specification Test Results

About this item

 :
 :136.09 g/mol
Merck Index:13,07744
Boiling Pt:∼100 °C (1013 hPa)
Melting Pt:253 °C
Density:2,3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-913-4
REACH:01-2119490224-41
 :GHS07
CAS Number:7778-77-0
Formula:KH₂PO₄
MDL Number:MFCD00011401