

Ethanol denatured, anhydrous 3C, Reagent Grade for biotechnology (denatured with 5% isopropanol)
Catalog # E402-4L
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:4 L
- Seq:5162
Specifications
About this item
:
:46.07 g/mol
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
:GHS02
:GHS07
CAS Number:64-17-5
Formula:H₃CCH₂OH
MDL Number:MFCD00003568