LAB-Q Ultra - Type I is a multipurpose water maker. This water maker produces Type I ultrapure water and Type II pure water simultaneously. The produced water has very low total organic carbon (TOC) content, meeting the requirements of liquid chromatography and all spectroscopy methods, besides microbiological and molecular biology application. It also meets ISO 3696, Type I Standards. Dispensing of ultrapure water is configured on the front panel whereas pre water is dispensed from the storage tank. For feed water with higher level of TDS i.e. >500 ppm a customised pre treatment module can be designed prior to INDION Lab-Q Ultra.
- Online resistivity meter- Displays product water quality for continuous monitoring
- Automatic flush device (AFD) – Ensures periodical cleaning of the filter system
- Integrated recirculation system – Ensures consistent quality of water and reduces TOC to low levels (2-5 ppb)
- Dry run protection – Ensures protection of the pump when the inlet water is unavailable
- Optional point-of-user filter – Ensure compliance to microbial reduction values
- Optional flexible dispensing – Remote water dispenser offers dispensing at various angles and heights
- Electrolytic auto sanitizer – Ensures periodic sanitization of upstream purifying filters
- High purity storage tank – A built-in UV, protective air filter and high grade tank material ensures sustained purity while in storage
Advantages: Works on tap water as sources; Compact and portable counter to model and Low operating cost which required minimum maintenance.
Applications include: Gas Chromatography, All spectroscopy methods, Enzyme immunoassays, HPLC (High performance liquid chromatography), ICP (Inductively coupled plasma), AAS (Atomic absorption spectrophotometer), Clinical research, Inorganic trace analysis, Buffer and media preparation, Molecular biology and Microbiological techniques.
- Catalog No:
- IONELQHE10012
- IONELQHE10011
- IONELQHU50012
- IONELQHP10011
- IONELQHU50011
- Model:
- Indion Lab Q Ultra Type 1 with 60 Liter Storage (EDI) Based
- Indion Lab Q Ultra Type 1 with 30 Liter Storage (EDI) Based
- Indion Lab Q Ultra Type 1 with 60 Liter Storage
- Indion Lab Q Polishing Unit
- Indion Lab Q Ultra Type 1 with 30 Liter Storage
- Feed water:
- TDS: 500 ppm Max..;
Fe, Mn: NIL;
Free Chlorine: NIL;
Silica: <20 ppm;
Inlet pressure: 0.5 - 2.0 kg/cm² - TDS: 500 ppm Max..;
Fe, Mn: NIL;
Free Chlorine: NIL;
Silica: <20 ppm;
Inlet pressure: 0.5 - 2.0 kg/cm² - TDS: 500 ppm Max..;
Fe, Mn: NIL;
Free Chlorine: NIL;
Silica: <20 ppm;
Inlet pressure: 0.5 - 2.0 kg/cm² - TDS: 500 ppm Max..;
Fe, Mn: NIL;
Free Chlorine: NIL;
Silica: <20 ppm;
Inlet pressure: 0.5 - 2.0 kg/cm² - TDS: 500 ppm Max..;
Fe, Mn: NIL;
Free Chlorine: NIL;
Silica: <20 ppm;
Inlet pressure: 0.5 - 2.0 kg/cm²
- Flow rate:
- 1 - 1.5 l/min, dispensing flow (10 L/h product flow to storage tank)
- 1 - 1.5 l/min, dispensing flow (10 L/h product flow to storage tank)
- 1 - 1.5 l/min, dispensing flow (10 L/h product flow to storage tank)
- 1 - 1.5 l/min, dispensing flow (10 L/h product flow to storage tank)
- 1 - 1.5 l/min, dispensing flow (10 L/h product flow to storage tank)
- Conductivity:
- 0,055 μS/cm (pure water conductivity - Type I);
<0,1 μS/cm (pure water conductivity - Type II) - 0,055 μS/cm (pure water conductivity - Type I);
<0,1 μS/cm (pure water conductivity - Type II) - 0,055 μS/cm (pure water conductivity - Type I);
<0,1 μS/cm (pure water conductivity - Type II) - 0,055 μS/cm (pure water conductivity - Type I);
<0,1 μS/cm (pure water conductivity - Type II) - 0,055 μS/cm (pure water conductivity - Type I);
<0,1 μS/cm (pure water conductivity - Type II)
- TOC content:
- <5 ppb
- <5 ppb
- <5 ppb
- <5 ppb
- <5 ppb
- Resistivity at +25 °C:
- 18.2 MΩ (pure water resisitivity Type I);
>10 MΩ (pure water resisitivity Type II); - 18.2 MΩ (pure water resisitivity Type I);
>10 MΩ (pure water resisitivity Type II); - 18.2 MΩ (pure water resisitivity Type I);
>10 MΩ (pure water resisitivity Type II); - 18.2 MΩ (pure water resisitivity Type I);
>10 MΩ (pure water resisitivity Type II); - 18.2 MΩ (pure water resisitivity Type I);
>10 MΩ (pure water resisitivity Type II);
- Bacteria:
- <0,1 cfu/ml
- <0,1 cfu/ml
- <0,1 cfu/ml
- <0,1 cfu/ml
- <0,1 cfu/ml
- Particles:
- >0,22 μm:<1 per ml
- >0,22 μm:<1 per ml
- >0,22 μm:<1 per ml
- >0,22 μm:<1 per ml
- >0,22 μm:<1 per ml
- Endotoxin:
- <0,001 EU/ml
- <0,001 EU/ml
- <0,001 EU/ml
- <0,001 EU/ml
- <0,001 EU/ml