About this item
INDION Lab-Q smart (Type II) water maker produces pure water which is essential for creating chemical reagents, microbiological buffers and media that are consistently pure. It is an excellent source of water for glassware washing and as feed water for laboratory equipment such as autoclaves, laboratory dishwashers and water baths. It is also a recommended source of pre-treated water for user in Type I ultrapure systems. It produces water with very low total organic carbon (TOC) content and conforms to ISO 3696 and corresponding ASTM standards. Portable tap water can be directly connected to INDION Lab-Q Smart to produce Type II pure water. It is easy to install and has several built-in safety features for reliable and consistent Type II water supply. For feed water with higher level of TDS i.e. >500 ppm a customised pre-treatment module can be designed prior to INDION Lab-Q Smart.
- Online resistivity meter – Displays product water quality for continuous monitoring
- Automatic flush device (AFD) – Ensure periodic cleaning of the system
- Dry run protection – Ensures protection of the pump when inlet water is unavailable
- Easy accessibility of components – All cartridges and filters are easily accessible and no special tool is required to replace them.
- Simple installation – Simple to install, operate and maintain with a clear indication of water purity
- Easy to dispense pure water
- Electrolytic auto sanitiser – Ensures periodic sanitisation for upstream purifying filters
Optional filtration: Filter can be attached at the point of dispensing spout to ensure compliance to microbial reduction values.
Optional flexible dispensing: Remote water dispenser offers dispensing at various angles and height.
Advantages: Tap water as source; compact and portable counter tap model ensures optional use of space; low operating cost and requires minimum maintenance.
Applications include flame spectrometry, electrochemistry, sample dilution, radioimmunoassay, electrophoresis, general chemistry, feed for Type I water maker, feed for clinical analysers, preparation of media and buffer, and autoclave and water bath.
Specifications
- Catalog No:
- IONELQHS20012
- IONELQHS20011
- Feed water:
- TDS: 500 ppm Max..;
Fe, Mn: NIL;
Free Chlorine: NIL;
Silica: <20 ppm;
Inlet pressure: 0.5 - 2.0 kg/cm² - TDS: 500 ppm Max..;
Fe, Mn: NIL;
Free Chlorine: NIL;
Silica: <20 ppm;
Inlet pressure: 0.5 - 2.0 kg/cm²
- Conductivity:
- <0,1 μS/cm
- <0,1 μS/cm
- TOC content:
- <30 ppb
- <30 ppb
- Resistivity at +25 °C:
- >10 MΩ
- >10 MΩ
- Bacteria:
- <1 cfu/ml
- <1 cfu/ml
- Particles:
- >0,22 μm:<1 per ml
- >0,22 μm:<1 per ml
- Endotoxin:
- <0,25 EU/ml
- <0,25 EU/ml