Order Entry
India
ContactUsLinkComponent
Penicillin G potassium salt
Irritant
Penicillin G potassium salt
Catalog # 441972W
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Penicillin G potassium salt
Catalog # 441972W
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :372.49 g/mol
Merck Index:12,07225
Melting Pt:214…217 °C
Density:1,359 g/cm³ (20 °C)
EINECS:204-038-0
 :GHS07
Storage Temperature:Refrigerator
CAS Number:113-98-4
Formula:C₁₆H₁₇KN₂O₄S
MDL Number:MFCD00036193