Order Entry
India
ContactUsLinkComponent
Iron(III) sulphate hydrate, AnalaR NORMAPUR® analytical reagent
 
 
Iron(III) sulphate hydrate, AnalaR NORMAPUR® analytical reagent
  24252.296
 :  
undefined
 
 
Iron(III) sulphate hydrate, AnalaR NORMAPUR® analytical reagent
  24252.296
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

 

 

  • Assay (calculated as Fe(III)):
    21.0 - 23.0 %
  • Insolubility in diluted HCl:
    Max. 100 ppm
  • Cl (Chloride):
    Max. 200 ppm
  • NO3 (Nitrate):
    Max. 0.5 %
  • PO4 (Phosphate):
    Max. 30 ppm
  • As (Arsenic):
    Max. 5 ppm
  • Cu (Copper):
    Max. 20 ppm
  • Fe (II) (Iron):
    Max. 0.02 %
  • Zn (Zinc):
    Max. 50 ppm
  • Ba (Barium):
    Max. 2 ppm
  • Mn (Manganese):
    Max. 0.1 %
  • Pb (Lead):
    Max. 6 ppm

 

 

 :
 :399.88 g/mol (anhy)
 :13,04065
 :3,1 g/cm³ (20 °C)
 :233-072-9
 :GHS07
 :GHS05
 :Ambient
 :15244-10-7
 :Fe₂(SO₄)₃·xH₂O
 :MFCD00149714