

Iron(III) sulphate hydrate, GPR RECTAPUR®
Catalog # 24248.292
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay (calculated as Fe(III)):Min. 21 %
- Cl (Chloride):Max. 100 ppm
- Fe (II) (Iron):Max. 0.05 %
- Zn (Zinc):Max. 100 ppm
Specification Test Results
About this item
:
:399.88 g/mol (anhy)
Merck Index:13,04065
Density:3,1 g/cm³ (20 °C)
EINECS:233-072-9
:GHS07
:GHS05
Storage Temperature:Ambient
CAS Number:15244-10-7
Formula:Fe₂(SO₄)₃·xH₂O
MDL Number:MFCD00149714