Order Entry
India
ContactUsLinkComponent
Sodium chloride 5 mol/L (5 M) in aqueous solution for molecular biology
Sodium chloride 5 mol/L (5 M) in aqueous solution for molecular biology
Catalog # A7006.1000
Supplier:  PanReac AppliChem
CAS Number:  
Sodium chloride 5 mol/L (5 M) in aqueous solution for molecular biology
Catalog # A7006.1000
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for liquids
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • DNases/RNases/Proteases:
    not detectable
  • Composition:
    NaCl: 292.2 g/L

Specification Test Results

About this item

 :
 :58.44 g/mol
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477