Solvent ASTM D2896, Chlorobenzene/Acetic acid-mixture (2/1 v/v), TBN solvent, VOLUSOL®
5154.2500
:
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
- Assay (Acetic acid):31.5 - 33.5 %
- Density (20/4):1.07 - 1.09
:
:>118 °C (1013 hPa)
:1,08 g/cm³ (20 °C)
:32 °C (closed cup)
:Ambient
:3265
:8,II
:GHS02
:GHS05
:GHS07
:GHS09