VWR®, Bovine serum albumin (BSA), Cohn fraction V
Catalog # 422371X
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 g
- CAS No.:9048-46-8
- Source:Serum
- Species:Bovine
- Storage conditions:Refrigerator
- Protein synonyms:BSA|Fraction V|Cohn Fraction V
- Protein/peptide name:Bovine Serum Albumin (BSA), Cohn Fraction V
- pH:pH 7
Specifications
About this item
A Cohn fraction V albumin produced by cold ethanol fractionation with minimum use of chemicals. Fatty acids and native sialic acid residues are preserved in this BSA.