Order Entry
India
ContactUsLinkComponent
2,6-Dichlorophenolindophenol sodium salt, tablets for the determination of vitamin C
2,6-Dichlorophenolindophenol sodium salt, tablets for the determination of vitamin C
Catalog # 230212X
Supplier:  VWR Chemicals
CAS Number:  
undefined
2,6-Dichlorophenolindophenol sodium salt, tablets for the determination of vitamin C
Catalog # 230212X
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    20 Tab.
  • Seq:
    0920
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Purple tablets
  • Shape:
    Biconvex
  • Tablet weight:
    114 to 140 mg
  • Diameter:
    6.0 - 7.0 mm
  • Average hardness (kp):
    Min. 2

Specification Test Results

About this item

Synonyms: Sodium 4-(3,5-dichloro-4-oxocyclohexa-2,5-dienylideneamino)phenoxide, Tillmans' reagent, 2,6-Dichloroindophenol sodium, 2,6-Dichloroindophenol sodium salt, 2,6-Dichloroindophenol sodium derivative
 :
 :290.08 g/mol
Merck Index:13,03093
Storage Temperature:Ambient
EINECS:210-640-4
CAS Number:620-45-1
Formula:C₁₂H₆Cl₂NNaO₂