Order Entry
India
ContactUsLinkComponent
VWR®, Pesticide reference standard, Performance Check Solution pesticides and PCBs, EPA 608
VWR®, Pesticide reference standard, Performance Check Solution pesticides and PCBs, EPA 608
Catalog # 124802M
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Pesticide reference standard, Performance Check Solution pesticides and PCBs, EPA 608
Catalog # 124802M
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pack type:
    Vial
  • Pk:
    1 mL
  • Description:
    Performance Check Solution pesticides and PCBs, EPA 608
Specifications
About this item
17 components.
20 µg/ml : Aldrin; a-BHC; b-BHC; d-BHC; g-BHC; p,p'-DDD; p,p'-DDE; Dieldrin; Endosulfan I; Endrin aldehyde; Heptachlor; Heptachlor epoxide (Isomer B); Methoxychlor
100 µg/ml : p,p'-DDT; Endosulfan II; Endosulfan sulfate; Endrin