Order Entry
India
ContactUsLinkComponent
Formic acid 98 - 100%, NORMATOM® for trace metal analysis
Formic acid 98 - 100%, NORMATOM® for trace metal analysis
  85905.270
 :  
undefined
Formic acid 98 - 100%, NORMATOM® for trace metal analysis
  85905.270
 :  

 

  • Pack type:
    Plastic bottle, with vented cap
  • Pk:
    500 mL
  • Seq:
    2725

 

 

  • Assay:
    Min. 98 %
  • Coloration:
    Max 10 APHA
  • Evaporation residue:
    Max. 0.0002 %
  • Cl (Chloride):
    Max. 1 ppm
  • Total S (as SO₄):
    Max. 1 ppm
  • Ag (Silver):
    Max 0.5 ppb
  • Al (Aluminium):
    Max 5 ppb
  • As (Arsenic):
    Max 1 ppb
  • Au (Gold):
    Max 0.5 ppb
  • Ba (Barium):
    Max 5 ppb
  • Be (Beryllium):
    Max 0.5 ppb
  • Bi (Bismuth):
    Max 0.5 ppb
  • Ca (Calcium):
    Max 10 ppb
  • Cd (Cadmium):
    Max 0.5 ppb
  • Co (Cobalt):
    Max 0.5 ppb
  • Cr (Chromium):
    Max 1 ppb
  • Cu (Copper):
    Max 1 ppb
  • Fe (Iron):
    Max 5 ppb
  • Ga (Gallium):
    Max 0.5 ppb
  • Ge (Germanium):
    Max 0.5 ppb
  • Hg (Mercury):
    Max 1 ppb
  • In (Indium):
    Max 0.5 ppb
  • K (Potassium):
    Max 5 ppb
  • Li (Lithium):
    Max 0.5 ppb
  • Mg (Magnesium):
    Max 5 ppb
  • Mn (Manganese):
    Max 0.5 ppb
  • Mo (Molybdenum):
    Max 0.5 ppb
  • Na (Sodium):
    Max 15 ppb
  • Ni (Nickel):
    Max 1 ppb
  • Pb (Lead):
    Max 1 ppb
  • Pd (Palladium):
    Max 1 ppb
  • Pt (Platinum):
    Max 1 ppb
  • Sb (Antimony):
    Max 1 ppb
  • Sn (Tin):
    Max 0.5 ppb
  • Sr (Strontium):
    Max 0.5 ppb
  • Ti (Titanium):
    Max 0.5 ppb
  • Tl (Thallium):
    Max 0.5 ppb
  • U (Uranium):
    Max 0.5 ppb
  • V (Vanadium):
    Max 0.5 ppb
  • Zn (Zinc):
    Max 5 ppb
  • Zr (Zirconium):
    Max 0.5 ppb

 

 

 :
 :46.03 g/mol
 :Ambient
 :1779
 :8 (3),II
 :GHS02
 :GHS05
 :64-18-6
 :HCO₂H
 :MFCD00003297