Order Entry
India
ContactUsLinkComponent
VWR®, Homogeniser, 4-Place Mini Bead Mill
VWR®, Homogeniser, 4-Place Mini Bead Mill
Catalog # VWRU10158-558
Supplier:  VWR Collection
VWR®, Homogeniser, 4-Place Mini Bead Mill
Catalog # VWRU10158-558
Supplier:  VWR Collection

Specifications

  • Capacity:
    25 µl - 5 ml
  • Description:
    4-Place Mini Bead Mill homogeniser
  • Electrical:
    100 – 230V, 50/60 Hz
  • Run time:
    1 sec to 5 min
  • Weight:
    7,48 kg
  • W×D×H:
    215×292×254 mm
  • Pk:
    1

Specifications

About this item

The VWR® 4-Place Mini Bead Mill Homogeniser is a compact and powerful bead mill homogenizer.

  • Specifically designed for grinding, lysing, and the homogenization of biological samples prior to molecular extraction
  • Ideal solution for releasing DNA, RNA, proteins, and enzymes from tough samples while still retaining molecular integrity
  • Offers a broad performance range

Using sample tubes which are pre-filled with a variety of lysing beads, the mill vigorously and uniformly shakes the tubes providing an efficient, consistent, high-yield and quality homogenisation in less than 60 seconds. Mini Bead Mill supports simultaneous processing of samples in 4×0,5 ml, 4×1,5 ml, 4×2 ml, or 1×7 ml tubes. It does not require a cool down period between runs, allowing non-stop use and high throughput. The unit features a convenient front loading design and safety lid closure.

Ordering information: Includes the VWR® Mini Bead Mill Drive Unit, integrated 0,5, 1,5 and 2 ml tube carriage, interchangeable 1x7 ml tube carriage and user manual. The VWR® disposable bead tubes must be ordered separately.