Order Entry
India
ContactUsLinkComponent
Methanol ≥99.90% for HPLC gradient analysis, Rankem®
Flammable
Toxic
Health
Methanol ≥99.90% for HPLC gradient analysis, Rankem®
Supplier:  RANKEM
Certificates
undefined
Flammable
Toxic
Health
Methanol ≥99.90% for HPLC gradient analysis, Rankem®
Supplier:  RANKEM
Certificates

Specification Test Results

  • Color (APHA):
    ≤ 10
  • Guarantee Analysis:
    .
  • Assay(GC):
    Min. 99.90 %
  • Weight Per ml (at 20°C):
    0.790 -- 0.792 g
  • Identity (IR):
    Conforms
  • Maximum Limits Of Impurities:
    .
  • Residue After Evaporation:
    Max. 0.0005 %
  • Titrable Acid:
    Max. 0.0002 meq/g
  • Titrable Base:
    Max. 0.0002 meq/g
  • Water ( By KF):
    Max. 0.03 %
  • UV Transmittance:
    .
  • Wavelength:
    .
  • 210 nm:
    Min. 30 %
  • 220 nm:
    Min. 60 %
  • 230 nm:
    Min. 80 %
  • 240 nm:
    Min. 90 %
  • From 260 nm:
    Min. 98 %
  • Gradient Performance:
    .
  • at 254 nm:
    ≤ 2 mAU
  • at 235 nm:
    ≤ 5 mAU

Specification Test Results

Frequently Bought Together