Order Entry
India
ContactUsLinkComponent
Hydrochloric acid solution N/10, Rankem®
Corrosive
Irritant
Hydrochloric acid solution N/10, Rankem®
Catalog # RANKH0210
Supplier:  RANKEM
CAS Number:  
Corrosive
Irritant
Hydrochloric acid solution N/10, Rankem®
Catalog # RANKH0210
Supplier:  RANKEM
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
ADR:8,I
 :36.46 g/mol
UN:1789
 :GHS05
 :GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324