Order Entry
India
ContactUsLinkComponent
n-Hexane (mixture of isomers) ≥98.50% for HPLC, Rankem®
Flammable
Health
Environment
n-Hexane (mixture of isomers) ≥98.50% for HPLC, Rankem®
Supplier:  RANKEM
Certificates
undefined
Flammable
Health
Environment
n-Hexane (mixture of isomers) ≥98.50% for HPLC, Rankem®
Supplier:  RANKEM
Certificates

Specification Test Results

  • Color (APHA):
    ≤ 10
  • Guarantee Analysis:
    .
  • Assay(GC)(Sum Of 5 Isomers And Methyl cyclopentane):
    ≥ 98.50 %
  • Weight Per ml (at 20°C):
    0.66 - 0.67 g
  • Identity (IR):
    Conforms
  • Maximum Limits Of Impurities:
    .
  • Residue After Evaporation:
    ≤ 0.0005 %
  • Titrable Acid:
    ≤ 0.0002 meq/g
  • Titrable Base:
    ≤ 0.0002 meq/g
  • Water ( By KF):
    ≤ 0.02 %
  • Sulphur Compounds.:
    <= 0.005%
  • Thiophene:
    Passes test
  • UV Transmittance:
    .
  • 200 nm:
    ≥ 20 %
  • 210 nm:
    ≥ 50 %
  • 220 nm:
    ≥ 80 %
  • 230 nm:
    ≥ 90 %
  • From 245 nm:
    ≥ 98 %

Specification Test Results

Related Information

Frequently Bought Together