

Hydrochloric acid min 35,00%, Laboratory Reagent, Rankem®
Supplier: RANKEM
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Density:1,18 g/cm³ (20 °C)
EINECS:231-595-7
UN:1789
ADR:8,II
:36.46 g/mol
:GHS05
:GHS07
Specification Test Results
- Assay (HCl):Min. 35.00%
- Specific Gravity At 25 ° C:1.178 --> 1.183
- Color:≤ 10 APHA
- Heavy Metals (As Pb):≤ 0.0005%
- Iron (Fe):≤ 0.0001%
- Residue On Ignition (SO₄²⁻):Max. 0.005 %
- Sulphate (SO₄²⁻):≤ 0.001%
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_RANKH0090 spec_RANKH0100 spec_RANKH0105 spec_RANKH0108
Product specification
Frequently Bought Together