Order Entry
India
ContactUsLinkComponent
Ethyl acetate ≥99.8% for HPLC, Rankem®
Flammable
Irritant
Ethyl acetate ≥99.8% for HPLC, Rankem®
Supplier:  RANKEM
Certificates
undefined
Flammable
Irritant
Ethyl acetate ≥99.8% for HPLC, Rankem®
Supplier:  RANKEM
Certificates

Specification Test Results

  • Colour (APHA):
    ≤10
  • Guarantee Analysis:
    .
  • Assay(GC):
    Min. 99.80%
  • Weight Per ml (at 20°C):
    0.900 --> 0.901 g
  • Identity (IR):
    Conforms
  • Maximum Limits Of Impurities:
    .
  • Residue After Evaporation:
    Max. 0.0005%
  • Titrable Acid:
    Max. 0.0009 meq/g
  • Water ( By KF):
    Max. 0.05%
  • UV Transmittance:
    .
  • Wavelength:
    .
  • 260 nm:
    Min. 50%
  • 265 nm:
    Min. 80%
  • From 275 nm:
    Min. 98%

Specification Test Results

Related Information

Frequently Bought Together