

Diethyl ether, DRYSOLV for organic synthesis, Rankem®
Supplier: RANKEM
Certificates
About this item
CAS Number:60-29-7
:
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646
:74.12 g/mol
UN:1155
ADR:3,I
:GHS02
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together