Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dichloromethane
Dichloromethane ≥99.9% for gas chromatography, Rankem®
Dichloromethane ≥99.9% for gas chromatography, Rankem®
RANKD0096
:
RANKEM
:
Dichloromethane ≥99.9% for gas chromatography, Rankem®
RANKD0096
:
RANKEM
:
Pk:
1 L
Seq:
5037
:
:
84.93 g/mol
:
39,8 °C (1013 hPa)
:
Ambient
:
1593
:
6.1,III
:
GHS07
:
GHS08
:
75-09-2
:
CH₂Cl₂
:
MFCD00000881