



2,2,4-Trimethylpentane ≥99.00% for HPLC, Rankem®
Supplier: RANKEM
Certificates
About this item
CAS Number:540-84-1
:
Formula:(CH₃)₂CHCH₂C(CH₃)₃
MDL Number:MFCD00008943
:114.23 g/mol
Boiling Pt:99,2 °C (1013 hPa)
Melting Pt:–107 °C
Density:0,692 g/cm³ (20 °C)
Flash Pt:−12 °C
EINECS:208-759-1
UN:1262
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Color (APHA):≤ 10
- Guarantee Analysis:.
- Assay(GC):Min. 99.00 %
- Weight Per ml (at 20°C):0.691 --> 0.692 g
- Identity (IR):Conforms
- Maximum Limits Of Impurities:.
- Residue After Evaporation:Max. 0.0005 %
- Titrable Acid:Max. 0.0002 meq/g
- Titrable Base:Max. 0.0002 meq/g
- Water ( By KF):Max. 0.01 %
- UV Transmittance:.
- Wavelength:.
- 210 nm:Min. 30 %
- 225 nm:Min. 80 %
- 235 nm:Min. 90 %
- From 250 nm:Min. 98 %
Specification Test Results
Related Information
Frequently Bought Together