Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
D(-)-Mannitol
D-(-)-Mannitol AR, Rankem®
D-(-)-Mannitol AR, Rankem®
D-(-)-Mannitol AR, Rankem®
Catalog # RANKM0294
Supplier:
RANKEM
CAS Number:
SDS
Certificates
Expand image
D-(-)-Mannitol AR, Rankem®
Catalog # RANKM0294
Supplier:
RANKEM
CAS Number:
SDS
Certificates
Specifications
Pk:
500 g
Seq:
3645
Specifications
About this item
Product Details
Documentation
:
:
182.17 g/mol
CAS Number:
69-65-8
Formula:
HOH₂C(CH(OH))₄CH₂OH
MDL Number:
MFCD00064287
Product Details
Documentation