


Mercury(II) bromide ≥99%, AnalaR NORMAPUR® ACS analytical reagent
86147.150
:
- Pk:50 g
- Seq:3368
- Assay:Min. 99 %
- Insoluble in methanol:Max. 0,05 %
- Residue after reduction:Max. 0,02 %
- Cl (Chloride):Max. 0,25 %
- Conforms to ACS:Passes test
:
:360.4 g/mol
:13,05900
:320 °C (1013 hPa)
:236 °C
:6,1 g/cm³ (20 °C)
:Ambient
:232-169-3
:1634
:6.1,II
:GHS06
:GHS08
:GHS09
:7789-47-1
:Br₂Hg
:MFCD00011040