Order Entry
India
ContactUsLinkComponent
Sodium dihydrogen phosphate monohydrate 98.0-103%, Multi-Compendial, Macron Fine Chemicals™
Sodium dihydrogen phosphate monohydrate 98.0-103%, Multi-Compendial, Macron Fine Chemicals™
  MACR7869-06
 :  
undefined
Sodium dihydrogen phosphate monohydrate 98.0-103%, Multi-Compendial, Macron Fine Chemicals™
  MACR7869-06
 :  

 

  • Pk:
    25 kg
  • Seq:
    4283

 

 

  • USP - Identification A :
    Passes test
  • USP - Identification B:
    Passes test
  • USP - pH:
    4.10 to 4.50
  • USP - Water:
    10.000 to 15.000 %
  • USP - Insoluble Substance :
    Max 0.2 %
  • USP - Chloride :
    Max 0.014 %
  • USP - Sulphate :
    Max. 0.15 %
  • USP - Aluminum,calcium,and related:
    Passes test
  • USP - Arsenic :
    Max 8 ppm
  • USP - Heavy Metal :
    0.002 % <0.002%
  • USP - Assay ( on anhydrous basis) :
    98.00 to 103.00 %
  • BP - Identification A:
    Passes test
  • BP - Identification B:
    Passes test
  • BP - Identification C:
    Passes test
  • BP - Identification D (LOD):
    11.5 to 14.5%
  • BP - Appearance of Solution:
    Passes test
  • BP - Acidity:
    4.20 to 4.50
  • BP - Reducing substances:
    Passes test
  • BP - Chloride :
    Max 200 ppm
  • BP - Sulphates :
    Max 300 ppm
  • BP - Arsenic :
    Max 2 ppm
  • BP - Iron :
    Max 10 ppm
  • BP - Heavy Metal :
    Max 10 ppm
  • BP - Loss on Drying:
    11.5 to 14.5%
  • BP - Assay (on the ignited basis ) :
    98.00 to 100.50 %

 

 

 :
 :137.99 g/mol
 :13,08734
 :100 °C
 :2,04 g/cm³
 :Ambient
 :231-449-2
 :01-2119489796-13
 :10049-21-5
 :NaH₂PO₄·H₂O
 :MFCD00149208