BeneSphera™ HIV Advanced ELISA Kit
Catalog # BENP15001002
Supplier: BENESPHERA
CAS Number:
Specifications
- Pk:96 Tests
- Description:BeneSphera™ HIV Advanced ELISA kit
Specifications
About this item
BeneSphera™ HIV Advanced ELISA kit is a fourth-generation immunoassay for qualitative detection of P24 antigen and antibodies to HIV-1 and HIV-2.
- Simultenious detection of antigen and antibodies to HIV 1 and 2
- Window period: 2 to 4 weeks
- Easy detection compared to 3rd generation assay
- Sensitivity: 100%
- Specificity: 100%
The window period for a 4th generation antigen/antibody test is about four weeks. By this time 95% of infections will be detected. There is a three-month window period after exposure, for the confirmatory result to detect more than 99.9% of infections.
Delivery: Kit includes: HIV ½ Antigen Coated Microstrips; Sample diluent; Control (positive/negative); Conjugate concentrate 101×; Conjugate diluent; TMB chromogen in DMSO; Substrate buffer; Washing solution concentrate (20×); Stop solution; Empty zip seal polybag; Plate sealer in zip seal polybag.
Caution: This product is designed for invitro diagnostic use only.