Order Entry
India
ContactUsLinkComponent
 
EDTA disodium salt dihydrate 99.0-101.0%, Multi-Compendial, Macron Fine Chemicals™
Catalog # MACRH410-06
CAS Number:  
undefined
EDTA disodium salt dihydrate 99.0-101.0%, Multi-Compendial, Macron Fine Chemicals™
Catalog # MACRH410-06
CAS Number:  

Specifications

  • Pk:
    25 L
  • Seq:
    5327

Specifications

Specification Test Results

  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Identification C:
    Passes Test
  • USP - pH (1 in 20):
    4.00 - 6.00
  • USP - Loss on Drying:
    8.7 - 11.4%
  • USP - Calcium (Ca):
    Passes Test
  • USP - Heavy Metals (as Pb):
    Max. 0.005%
  • USP - Nitrilotriacetic Acid:
    Max. 0.1%
  • USP - Assay (C₁₀H₁₄N₂Na₂O₈) Dried:
    99.00 - 101.00%
  • USP - Endotoxin Concentration (EU/g):
    Max.5
  • BP/EP - Assay (C₁₀H₁₄N₂Na₂O₈·2H₂O):
    98.50 - 101.00%
  • BP/EP - Identification A:
    Passes Test
  • BP/EP - Identification B:
    Passes Test
  • BP/EP - Identification C:
    Passes Test
  • BP/EP - Identification D:
    Passes Test
  • BP/EP - pH (1 in 20):
    4.00 - 5.50
  • BP/EP - Heavy Metals (as Pb):
    Max. 20 ppm
  • BP/EP - Iron (Fe):
    Max. 80 ppm
  • BP/EP - Impurity A:
    Max. 0.1%
  • JP - Assay (C₁₀H₁₄N₂Na₂O₈) Dried:
    99.00 - 101.00%
  • JP - Identification A:
    Passes Test
  • JP - Identification B:
    Passes Test
  • JP - Identification C:
    Passes Test
  • JP - pH of 1% Solution at 25 °C:
    4.30 - 4.70
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Cyanide (CN):
    Passes Test
  • JP - Heavy Metals (as Pb):
    Max. 10 ppm
  • JP - Arsenic (As):
    Max. 2 ppm
  • JP - Residue on Ignition:
    37.0 - 39.0%

Specification Test Results

About this item

 :
 :372.24 g/mol
CAS Number:6381-92-6
Formula:C₁₀H₁₄N₂Na₂O₈·2H₂O
MDL Number:MFCD00150037