Order Entry
India
ContactUsLinkComponent
VWR®, Bottle, Round Bottom, with Thread
VWR®, Bottle, Round Bottom, with Thread
Catalog # 548-1980
Supplier:  VWR Collection
VWR®, Bottle, Round Bottom, with Thread
Catalog # 548-1980
Supplier:  VWR Collection

Specifications

  • Capacity:
    20 ml
  • Colour:
    Clear
  • Description:
    Fine thread bottle, 75,5×22,5 mm
  • Pk:
    1.000

Specifications

About this item

These round bottom bottles are made up of first class hydrolytic grade.

  • With fill marks
  • Magentic screw caps