


Methanol ≥99.50% (by GC), Multi-Compendial, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:67-56-1
:
Formula:CH₃OH
MDL Number:MFCD00004595
:32.04 g/mol
Storage Temperature:Ambient
UN:1230
ADR:3,II
REACH:01-2119433307-44
:GHS02
:GHS06
:GHS08
Specification Test Results
- Assay (CH₃OH) (By GC):≥99.50%
- Identification A:Passes Test
- Identification B:Passes Test
- Acidity:Passes Test
- Alkalinity (As NH₃):Max. 3 ppm
- Non Volatile Residue:≤0.0010%
- Readialy Carbonisable Substances:Passes Test
- Readialy Oxidisable Substances:Passes Test
- Acetone ,Aldehydes (As(CH₃)2CO):Max. 0.003%
- Water (H2O) (By KF):≤0.100%
- Appearance:Passes test
- Acidity Or Alkalinity:Passes test
- Relative Density:0.7910 - 0.7930
- Absorbance, 230 nm:≤0.1500
- Absorbance, 250 nm:≤0.0500
- Absorbance, 270 nm:≤0.0200
- Absorbance, 290 nm:≤0.0100
- Absorbance, 230 - 290 nm:Passes test
- Impurity A (Benzene):Max 2 ppm
- Single Impurity Peak:max 0.1%
- Total Of All Impurity Peaks:Max 0.3%
- Reducing Substances:Passes test
- Residue On Evaporation:Max 10 ppm
- Water:≤0.100%
Specification Test Results
Frequently Bought Together