Order Entry
India
ContactUsLinkComponent
VWR®, Glycylglycine (Gly-Gly)
VWR®, Glycylglycine (Gly-Gly)
Catalog # VWRA0137-5KG
Supplier:  VWR Collection
CAS Number:  
VWR®, Glycylglycine (Gly-Gly)
Catalog # VWRA0137-5KG
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pk:
    5 kg
  • CAS No.:
    556-50-3
  • Storage conditions:
    Ambient
  • MDL No.:
    MFCD00008130
  • Protein synonyms:
    Glycylglycine-Gly-Gly
  • Protein/peptide name:
    Glycylglycine (Gly-Gly)
  • Purity:
    99,0%
  • Molecular weight:
    132,12 g/mol
  • Grade:
    High purity grade
  • Molecular formula:
    C₄H₈N₂O₃

Specifications

About this item

Glycylglycine (Gly-Gly) contain ammonia (NH₃) (Max.002%), arsenic (Max. 0,0001%), ash (Max. 0,2%), chloride (Max. 0,02%), heavy metals (As Pb) (Max. 0,002%), iron (Max. 0,001%) and sulphate (Max. 0,03%).

  • Identification(By IR): Pass
  • Loss On drying: Max. 0,2%
  • Solubility (10%, water): Pass