Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tert-Butanol
tert-Butanol AR, Rankem®
tert-Butanol AR, Rankem®
tert-Butanol AR, Rankem®
Catalog # RANKB0421
Supplier:
RANKEM
CAS Number:
Certificates
Expand image
tert-Butanol AR, Rankem®
Catalog # RANKB0421
Supplier:
RANKEM
CAS Number:
Certificates
Specifications
Pk:
500 mL
Seq:
2725
Specifications
About this item
Product Details
Documentation
:
:
74.12 g/mol
CAS Number:
75-65-0
Formula:
(CH₃)₃COH
MDL Number:
MFCD00004464
Product Details
Documentation