Acetonitrile ≥98.9% for HPLC gradient analysis, Rankem®
Supplier: RANKEM
Certificates
About this item
CAS Number:75-05-8
:
Formula:H₃CCN
MDL Number:MFCD00001878
:41.05 g/mol
Boiling Pt:81,6 °C (1013 hPa)
Melting Pt:–45,7 °C
Density:0,782 g/cm³ (20 °C)
Flash Pt:2 °C (closed cup)
Storage Temperature:Ambient
UN:1648
ADR:3,II
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Guarantee Analysis:.
- Assay(GC):Min. 99.90 %
- Weight Per ml (at 20°C):0.782 -- 0.783 g
- Maximum Limits Of Impurities:.
- Acidity:Max. 0.0002 meq/g
- Alkalinity:Max. 0.0002 meq/g
- Non volatile matter:Max. 0.0004 %
- Water ( By KF):Max. 0.03 %
- UV Transmittance:.
- Wavelength:.
- 195 nm:Min. 75 %
- 200 nm:Min. 93 %
- 210 nm:Min. 96 %
- 220 nm:Min. 97 %
- from 230 nm:Min. 98 %
- Gradient Performance:.
- at 254 nm:≤ 1 mAU
- at 210 nm:≤ 5 mAU
Specification Test Results
Related Information
Frequently Bought Together