Dextrose, anhydrous, Multi-Compendial, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:50-99-7
:492-62-6
Formula:C₆H₁₂O₆
MDL Number:MFCD00148912
:180.16 g/mol
Specification Test Results
- USP - Identification:Passes Test
- USP - Specific Rotation áD₂₅, (Calculated on Dried Basis):+52.60 to +53.20°
- USP - Colour of Solution:Passes Test
- USP-Acidity:Passes Test
- USP-Water (H2O):Max. 0.5%
- USP - Residue on Ignition:Max. 0.1%
- USP - Chloride (Cl):Max. 0.018%
- USP - Sulphate (SO4):Max. 0.025%
- USP - Arsenic (As):Max. 1 ppm
- USP - Heavy Metals (as Pb):Max. 5 ppm
- USP - Dextrin:Passes Test
- USP - Soluble Starch, Sulphites:Passes Test
- USP - Endotoxin Concentration (EU/g:<2.5
- BP/EP - Identification A:Passes Test
- BP/EP - Identification B:Passes Test
- BP/EP - Identification C:Passes Test
- BP/EP - Appearance of Solution:Passes Test
- BP/EP - Acidity or Alkalinity:Passes Test
- BP/EP -Specific Rotation áD₂₀. (Calculated on Dried Basis):+52.50 to +53.30°
- BP/EP - Foreign Sugars, Soluble Starch,and Dextrins:Passes Test
- BP/EP - Sulphite(SO₂):Max. 15 ppm
- BP/EP - Chloride (Cl):Max. 125 ppm
- BP/EP - Sulfate (SO₄):Max.200 ppm
- BP/EP - Arsenic (As):Max. 1 ppm
- BP/EP - Barium (Ba):Passes Test
- BP/EP - Calcium (Ca):Max. 200 ppm
- BP/EP - Lead (Pb):Max. 0.5 ppm
- BP/EP - Water (H2O):Max. 1.0%
- BP/EP - Sulphated Ash:Max. 0.1%
- JP - Assay(dried basis):99.50 to 101.00%
- JP - Identification A:Passes Test
- JP - Clarity and Colour of Solution:Passes Test
- JP - Acidity or Alkalinity:Passes Test
- JP - Chloride (Cl):Max. 0.018%
- JP - Sulfates (SO4):Max. 0.024%
- JP - Heavy Metals (as Pb):Max. 4 ppm
- JP - Arsenic (As):Max. 1.3 ppm
- JP - Dextrin:Passes Test
- JP - Soluble Starch, Sulfites:Passes Test
- JP - Loss on Drying at 105 °C:Max. 1.0%
- JP - Residue on Ignition:Max. 0.10%
Specification Test Results
Frequently Bought Together