


2-Propanol ≥99.5% (by GC, corrected for water content), CMOS for disk drives, J.T.Baker®
Catalog # 9078-05
Supplier: Avantor
CAS Number:
Specifications
- Pk:4 L
- Seq:5162
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Assay (CH₃CHOHCH₃) (by GC, corrected for water):≥ 99.5 %
- Color (APHA):≤ 10
- Residue after Evaporation:≤ 4 ppm
- Solubility in H₂O:Passes Test
- Water (H₂O)(by Karl Fischer titrn):≤ 0.05 %
- Acidity (µeq/g):≤ 0.2
- Alkalinity (µeq/g):≤ 0.1
- Silicone(Absorbance at 1050 cm -1):Passes Test
- Phthalate Esters(by FT-IR)(0.1 A.U. Max. at 1700 - 1750 cm-1):Passes Test
- Chloride (Cl):≤ 0.2 ppm
- Phosphate (PO₄):≤ 0.3 ppm
- Trace Impurities - Aluminum (Al):≤ 50.0 ppb
- Arsenic and Antimony (as As):≤ 10.0 ppb
- Trace Impurities - Barium (Ba):≤ 20.0 ppb
- Trace Impurities - Boron (B):≤ 10.0 ppb
- Trace Impurities - Cadmium (Cd):≤ 20.0 ppb
- Trace Impurities - Calcium (Ca):≤ 50.0 ppb
- Trace Impurities - Chromium (Cr):≤ 20.0 ppb
- Trace Impurities - Cobalt (Co):≤ 20 ppb
- Trace Impurities - Copper (Cu):≤ 10.0 ppb
- Trace Impurities - Gallium (Ga):≤ 50 ppb
- Trace Impurities - Germanium (Ge):≤ 50.0 ppb
- Trace Impurities - Gold (Au):≤ 20 ppb
- Heavy Metals (as Pb):≤ 200.0 ppb
- Trace Impurities - Iron (Fe):≤ 50.0 ppb
- Trace Impurities - Lead (Pb):≤ 20.0 ppb
- Trace Impurities - Lithium (Li):≤ 50.0 ppb
- Trace Impurities - Magnesium (Mg):≤ 20 ppb
- Trace Impurities - Manganese (Mn):≤ 20.0 ppb
- Trace Impurities - Molybdenum (Mo):≤ 300.0 ppb
- Trace Impurities - Nickel (Ni):≤ 10.0 ppb
- Trace Impurities - Potassium (K):≤ 100 ppb
- Trace Impurities - Silicon (Si):≤ 50 ppb
- Trace Impurities - Silver (Ag):≤ 20.0 ppb
- Trace Impurities - Sodium (Na):≤ 100.0 ppb
- Trace Impurities - Strontium (Sr):≤ 20.0 ppb
- Trace Impurities - Tin (Sn):≤ 100 ppb
- Trace Impurities - Titanium (Ti):≤ 20 ppb
- Trace Impurities - Vanadium (V):≤ 300.0 ppb
- Trace Impurities - Zinc (Zn):≤ 50 ppb
- Trace Impurities - Zirconium (Zr):≤ 300.0 ppb
- Particle Count - 1.0 µm and greater:≤ 10 par/ml
Specification Test Results
About this item
:
:60.1 g/mol
ADR:3,II
:Management of Change (MOC) category = R
UN:1219
:GHS02
:GHS07
:GHS08
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674